missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 4-Fluorobenzoate 98.0+%, TCI America™
$321.51 - $321.51
Chemical Identifiers
| CAS | 451-46-7 |
|---|---|
| Molecular Formula | C9H9FO2 |
| Molecular Weight (g/mol) | 168.167 |
| MDL Number | MFCD00000351 |
| InChI Key | UMPRJGKLMUDRHL-UHFFFAOYSA-N |
| Synonym | 4-fluorobenzoic acid ethyl ester, ethyl p-fluorobenzoate, benzoic acid, 4-fluoro-, ethyl ester, p-fluorobenzoic acid, ethyl ester, ethyl4-fluorobenzoate, 4-fluoro-benzoic acid ethyl ester, benzoic acid, p-fluoro-, ethyl ester, ethyep-fluorobenzoate, pubchem3471, ethyl-4-fluorobenzoate |
| PubChem CID | 67976 |
| IUPAC Name | ethyl 4-fluorobenzoate |
| SMILES | CCOC(=O)C1=CC=C(C=C1)F |
Chemical Identifiers
| 451-46-7 | |
| 168.167 | |
| UMPRJGKLMUDRHL-UHFFFAOYSA-N | |
| 67976 | |
| CCOC(=O)C1=CC=C(C=C1)F |
| C9H9FO2 | |
| MFCD00000351 | |
| 4-fluorobenzoic acid ethyl ester, ethyl p-fluorobenzoate, benzoic acid, 4-fluoro-, ethyl ester, p-fluorobenzoic acid, ethyl ester, ethyl4-fluorobenzoate, 4-fluoro-benzoic acid ethyl ester, benzoic acid, p-fluoro-, ethyl ester, ethyep-fluorobenzoate, pubchem3471, ethyl-4-fluorobenzoate | |
| ethyl 4-fluorobenzoate |
Specifications
| 451-46-7 | |
| 210°C | |
| MFCD00000351 | |
| 4-fluorobenzoic acid ethyl ester, ethyl p-fluorobenzoate, benzoic acid, 4-fluoro-, ethyl ester, p-fluorobenzoic acid, ethyl ester, ethyl4-fluorobenzoate, 4-fluoro-benzoic acid ethyl ester, benzoic acid, p-fluoro-, ethyl ester, ethyep-fluorobenzoate, pubchem3471, ethyl-4-fluorobenzoate | |
| CCOC(=O)C1=CC=C(C=C1)F | |
| 168.167 | |
| 168.17 | |
| Crystalline Lumps |
| White | |
| C9H9FO2 | |
| 25 g | |
| UMPRJGKLMUDRHL-UHFFFAOYSA-N | |
| ethyl 4-fluorobenzoate | |
| 67976 | |
| ≥98.0% (GC) | |
| Ethyl 4-Fluorobenzoate |
Safety and Handling
TSCA : Yes