missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 4-Fluorobenzoate 98.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Boiling Point | 210°C |
| Quantity | 25 g |
| Formula Weight | 168.17 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Lumps |
| Chemical Name or Material | Ethyl 4-Fluorobenzoate |
Safety and Handling
| TSCA | Yes |