missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-Ethoxybenzoate 97.0+%, TCI America™

Catalog No. E052425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E052425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E052425G Supplier TCI America Supplier No. E052425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 23676-09-7
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00009116
InChI Key HRAQMGWTPNOILP-UHFFFAOYSA-N
Synonym benzoic acid, 4-ethoxy-, ethyl ester, 4-ethoxybenzoic acid ethyl ester, 4-ethoxy ethylbenzoate, benzoic acid, p-ethoxy-, ethyl ester, 4-ethoxyethylbenzoate, peeb, ethyl p-ethoxybenzoate, ethyl-4-ethoxybenzoate, p-ethoxy ethyl benzoate, ethyl-p-ethoxy benzoate
PubChem CID 90232
IUPAC Name ethyl 4-ethoxybenzoate
SMILES CCOC1=CC=C(C=C1)C(=O)OCC

Specifications

Color Yellow
Boiling Point 275°C
Quantity 25 g
Formula Weight 194.23
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 4-Ethoxybenzoate