missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-ethoxybenzoate, 98%

Catalog No. p-7022301
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1501014 25 g
AAA1501022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1501014 Supplier Thermo Scientific Chemicals Supplier No. A1501014

CAS: 23676-09-7 | C11H14O3 | 194.23 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.

Chemical Identifiers

CAS 23676-09-7
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00009116
InChI Key HRAQMGWTPNOILP-UHFFFAOYSA-N
Synonym benzoic acid, 4-ethoxy-, ethyl ester, 4-ethoxybenzoic acid ethyl ester, 4-ethoxy ethylbenzoate, benzoic acid, p-ethoxy-, ethyl ester, 4-ethoxyethylbenzoate, peeb, ethyl p-ethoxybenzoate, ethyl-4-ethoxybenzoate, p-ethoxy ethyl benzoate, ethyl-p-ethoxy benzoate
PubChem CID 90232
IUPAC Name ethyl 4-ethoxybenzoate
SMILES CCOC1=CC=C(C=C1)C(=O)OCC

Specifications

Melting Point 9°C
Density 1.071
Boiling Point 145°C to 150°C (13 mmHg)
Flash Point 137°C (278°F)
Refractive Index 1.518
Quantity 25 g
Beilstein 2692236
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 194.23
Percent Purity 98%
Chemical Name or Material Ethyl 4-ethoxybenzoate
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 245-818-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only