missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-Cyanocinnamate 98.0+%, TCI America™

Catalog No. E06775G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E06775G 5 g
E06771G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E06775G Supplier TCI America Supplier No. E06775G

Chemical Identifiers

CAS 62174-99-6
Molecular Formula C12H11NO2
Molecular Weight (g/mol) 201.225
InChI Key NVLBOCIUMFYPGI-BQYQJAHWSA-N
Synonym 4-Cyanocinnamic Acid Ethyl Ester
PubChem CID 5373497
IUPAC Name ethyl (E)-3-(4-cyanophenyl)prop-2-enoate
SMILES CCOC(=O)C=CC1=CC=C(C=C1)C#N

Specifications

Melting Point 70°C
Quantity 5 g
UN Number 3439
Formula Weight 201.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 4-Cyanocinnamate