accessibility menu, dialog, popup

UserName

H3COON(E/Z)

CAS RN 62174-99-6

IUPAC Name: ethyl 3-(4-cyanophenyl)prop-2-enoate
Synonyms: ethyl 3-(4-cyanophenyl)prop-2-enoate
Molecular Weight (g/mol): 201.23
Molecular Formula: C12H11NO2
InChi Key: NVLBOCIUMFYPGI-UHFFFAOYSA-N
SMILES: CCOC(=O)C=CC1=CC=C(C=C1)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results