missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-Chlorobenzoate 96.0+%, TCI America™

Catalog No. C162425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C162425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C162425G Supplier TCI America Supplier No. C162425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7335-27-5
Molecular Formula C9H9ClO2
Molecular Weight (g/mol) 184.62
MDL Number MFCD00013645
InChI Key RWBYCMPOFNRISR-UHFFFAOYSA-N
Synonym ethyl p-chlorobenzoate, benzoic acid, 4-chloro-, ethyl ester, ethyl-4-chlorobenzoate, p-ethoxycarbonyl phenyl chloride, ethyl4-chlorobenzoate, rarechem al bi 0077, 4-chlorobenzoic acid ethyl ester, ethyl p-chlorobenzate, benzoic acid, p-chloro-, ethyl ester, pubchem3705
PubChem CID 81785
IUPAC Name ethyl 4-chlorobenzoate
SMILES CCOC(=O)C1=CC=C(Cl)C=C1

Specifications

Color Yellow
Boiling Point 238°C
Quantity 25 g
Formula Weight 184.62
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 4-Chlorobenzoate