missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 4-chlorobenzoate, 98+%
CAS: 7335-27-5 | C9H9ClO2 | 184.62 g/mol
$102.74 - $411.32
Chemical Identifiers
| CAS | 7335-27-5 |
|---|---|
| Molecular Formula | C9H9ClO2 |
| Molecular Weight (g/mol) | 184.62 |
| MDL Number | MFCD00013645 |
| InChI Key | RWBYCMPOFNRISR-UHFFFAOYSA-N |
| Synonym | ethyl p-chlorobenzoate, benzoic acid, 4-chloro-, ethyl ester, ethyl-4-chlorobenzoate, p-ethoxycarbonyl phenyl chloride, ethyl4-chlorobenzoate, rarechem al bi 0077, 4-chlorobenzoic acid ethyl ester, ethyl p-chlorobenzate, benzoic acid, p-chloro-, ethyl ester, pubchem3705 |
| PubChem CID | 81785 |
| IUPAC Name | ethyl 4-chlorobenzoate |
| SMILES | CCOC(=O)C1=CC=C(Cl)C=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1254018
|
Thermo Scientific Chemicals
A1254018 |
50 g |
Each for $102.74
|
|
|||||
|
AAA1254030
|
Thermo Scientific Chemicals
A1254030 |
250 g |
Each for $411.32
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7335-27-5 | |
| 184.62 | |
| RWBYCMPOFNRISR-UHFFFAOYSA-N | |
| 81785 | |
| CCOC(=O)C1=CC=C(Cl)C=C1 |
| C9H9ClO2 | |
| MFCD00013645 | |
| ethyl p-chlorobenzoate, benzoic acid, 4-chloro-, ethyl ester, ethyl-4-chlorobenzoate, p-ethoxycarbonyl phenyl chloride, ethyl4-chlorobenzoate, rarechem al bi 0077, 4-chlorobenzoic acid ethyl ester, ethyl p-chlorobenzate, benzoic acid, p-chloro-, ethyl ester, pubchem3705 | |
| ethyl 4-chlorobenzoate |
Specifications
| 7335-27-5 | |
| 237°C to 239°C | |
| 1.524 | |
| 50 g | |
| ethyl p-chlorobenzoate, benzoic acid, 4-chloro-, ethyl ester, ethyl-4-chlorobenzoate, p-ethoxycarbonyl phenyl chloride, ethyl4-chlorobenzoate, rarechem al bi 0077, 4-chlorobenzoic acid ethyl ester, ethyl p-chlorobenzate, benzoic acid, p-chloro-, ethyl ester, pubchem3705 | |
| CCOC(=O)C1=CC=C(Cl)C=C1 | |
| 184.62 | |
| 184.62 | |
| Ethyl 4-chlorobenzoate |
| 1.18 | |
| C9H9ClO2 | |
| MFCD00013645 | |
| 2045882 | |
| RWBYCMPOFNRISR-UHFFFAOYSA-N | |
| ethyl 4-chlorobenzoate | |
| 81785 | |
| ≥98% |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 230-844-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only