missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-chlorobenzoate, 98+%

Catalog No. AAA1254018
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1254018 50 g
AAA1254030 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1254018 Supplier Thermo Scientific Chemicals Supplier No. A1254018
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 7335-27-5 | C9H9ClO2 | 184.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7335-27-5
Molecular Formula C9H9ClO2
Molecular Weight (g/mol) 184.62
MDL Number MFCD00013645
InChI Key RWBYCMPOFNRISR-UHFFFAOYSA-N
Synonym ethyl p-chlorobenzoate, benzoic acid, 4-chloro-, ethyl ester, ethyl-4-chlorobenzoate, p-ethoxycarbonyl phenyl chloride, ethyl4-chlorobenzoate, rarechem al bi 0077, 4-chlorobenzoic acid ethyl ester, ethyl p-chlorobenzate, benzoic acid, p-chloro-, ethyl ester, pubchem3705
PubChem CID 81785
IUPAC Name ethyl 4-chlorobenzoate
SMILES CCOC(=O)C1=CC=C(Cl)C=C1

Specifications

Density 1.18
Boiling Point 237°C to 239°C
Refractive Index 1.524
Quantity 50 g
Beilstein 2045882
Formula Weight 184.62
Percent Purity ≥98%
Chemical Name or Material Ethyl 4-chlorobenzoate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 230-844-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only