missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 4-chloro-8-cyanoquinoline-3-carboxylate, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 449680050
| Quantity | 5g |
|---|
Chemical Identifiers
| 77173-67-2 | |
| 260.68 | |
| ethyl4-chloro-8-cyanoquinoline-3-carboxylate, ethyl 4-chloro-8-cyano-3-quinolinecarboxylate, pubchem19877, 4-chloro-8-cyanoquinoline-3-carboxylicacidethyleste, 4-chloro-8-cyanoquinoline-3-carboxylic acid ethyl ester, ethyl 4-chloro-8-cyanoquinoxaline-3-carboxylate | |
| ethyl 4-chloro-8-cyanoquinoline-3-carboxylate |
| C13H9ClN2O2 | |
| MHOFAUCLSGVKOH-UHFFFAOYSA-N | |
| 3761179 | |
| CCOC(=O)C1=C(C2=CC=CC(=C2N=C1)C#N)Cl |
Specifications
| Ethyl 4-chloro-8-cyanoquinoline-3-carboxylate | |
| Authentic | |
| C13H9ClN2O2 | |
| ethyl4-chloro-8-cyanoquinoline-3-carboxylate, ethyl 4-chloro-8-cyano-3-quinolinecarboxylate, pubchem19877, 4-chloro-8-cyanoquinoline-3-carboxylicacidethyleste, 4-chloro-8-cyanoquinoline-3-carboxylic acid ethyl ester, ethyl 4-chloro-8-cyanoquinoxaline-3-carboxylate | |
| CCOC(=O)C1=C(C2=CC=CC(=C2N=C1)C#N)Cl | |
| 260.68 | |
| 260.68 |
| 77173-67-2 | |
| 96% min. (HPLC) | |
| 5g | |
| MHOFAUCLSGVKOH-UHFFFAOYSA-N | |
| ethyl 4-chloro-8-cyanoquinoline-3-carboxylate | |
| 3761179 | |
| 97% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning