missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-chloro-8-cyanoquinoline-3-carboxylate, 97%, Thermo Scientific™

Catalog No. p-7077329
Change view
Click to view available options
Quantity:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC449680010 1g
AC449680050 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC449680010 Supplier Thermo Scientific Chemicals Supplier No. 449680010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 77173-67-2
Molecular Formula C13H9ClN2O2
Molecular Weight (g/mol) 260.68
InChI Key MHOFAUCLSGVKOH-UHFFFAOYSA-N
Synonym ethyl4-chloro-8-cyanoquinoline-3-carboxylate, ethyl 4-chloro-8-cyano-3-quinolinecarboxylate, pubchem19877, 4-chloro-8-cyanoquinoline-3-carboxylicacidethyleste, 4-chloro-8-cyanoquinoline-3-carboxylic acid ethyl ester, ethyl 4-chloro-8-cyanoquinoxaline-3-carboxylate
PubChem CID 3761179
IUPAC Name ethyl 4-chloro-8-cyanoquinoline-3-carboxylate
SMILES CCOC(=O)C1=C(C2=CC=CC(=C2N=C1)C#N)Cl

Specifications

Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1g
Formula Weight 260.68
Percent Purity 97%
Chemical Name or Material Ethyl 4-chloro-8-cyanoquinoline-3-carboxylate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

RUO – Research Use Only