missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-chloro-6-hydroxy-5-nitronicotinate, 97%, Thermo Scientific™

Catalog No. p-7052432
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3319203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3319203 Supplier Thermo Scientific Chemicals Supplier No. H3319203

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Notes
Incompatible with strong oxidizing agents and strong acids.

Chemical Identifiers

CAS 1210835-74-7
Molecular Formula C8H7ClN2O5
Molecular Weight (g/mol) 246.603
MDL Number MFCD22989339
InChI Key FETZVQJUBMRFJE-UHFFFAOYSA-N
Synonym ethyl 4-chloro-5-nitro-6-oxo-1,6-dihydropyridine-3-carboxylate, ethyl 4-chloro-6-hydroxy-5-nitronicotinate, ethyl 4-chloro-6-hydroxy-5-nitropyridine-3-carboxylate, ethylchloronitrooxodihydropyridinecarboxylate, 4-chloro-6-hydroxy-5-nitronicotinic acid ethyl ester, ethyl 4-chloro-5-nitro-6-oxo-1,6-dihydro-3-pyridinecarboxylate, 4-chloro-5-nitro-6-oxo-1,6-dihydro-pyridine-3-carboxylic acid ethyl ester
PubChem CID 59225818
IUPAC Name ethyl 4-chloro-5-nitro-6-oxo-1H-pyridine-3-carboxylate
SMILES CCOC(=O)C1=CNC(=O)C(=C1Cl)[N+](=O)[O-]

Specifications

Melting Point 176°C to 178°C
Color Pink to White
Quantity 1 g
Formula Weight 246.6
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Ethyl 4-chloro-6-hydroxy-5-nitronicotinate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures