accessibility menu, dialog, popup

UserName

CAS RN 1210835-74-7

IUPAC Name: ethyl 4-chloro-5-nitro-6-oxo-1,6-dihydropyridine-3-carboxylate
Synonyms: ethyl 4-chloro-5-nitro-6-oxo-1H-pyridine-3-carboxylate
Molecular Weight (g/mol): 246.6
Molecular Formula: C8H7ClN2O5
InChi Key: FETZVQJUBMRFJE-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CNC(=O)C(=C1Cl)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results