missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 4-Chloro-2-(methylthio)pyrimidine-5-carboxylate 97.0+%, TCI America™
Supplier: TCI America E084125G
Specifications
| Ethyl 4-Chloro-2-(methylthio)pyrimidine-5-carboxylate | |
| 62°C | |
| 132°C | |
| MFCD00006085 | |
| ethyl 4-chloro-2-methylthio pyrimidine-5-carboxylate, ethyl 4-chloro-2-methylthio-5-pyrimidinecarboxylate, ethyl 4-chloro-2-methylthiopyrimidine-5-carboxylate, 4-chloro-2-methylsulfanyl-pyrimidine-5-carboxylic acid ethyl ester, ethyl 4-chloro-2-methylsulfanyl pyrimidine-5-carboxylate, 2-methylthio-4-chloro-5-ethoxycarbonylpyrimidine, 4-chloro-5-carbethoxy-2-methylthiopyrimidine, 5-pyrimidinecarboxylic acid, 4-chloro-2-methylthio-, ethyl ester, 4-chloro-2-methylthio pyrimidine-5-carboxylic acid ethyl ester, 4-chloro-2-methylsulfanylpyrimidine-5-carboxylic acid ethyl ester | |
| CCOC(=O)C1=CN=C(N=C1Cl)SC | |
| 232.682 | |
| 232.68 | |
| Crystalline Powder |
| 5909-24-0 | |
| White-Yellow | |
| C8H9ClN2O2S | |
| 25 g | |
| SNNHLSHDDGJVDM-UHFFFAOYSA-N | |
| ethyl 4-chloro-2-methylsulfanylpyrimidine-5-carboxylate | |
| 80008 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 5909-24-0 | |
| 232.682 | |
| SNNHLSHDDGJVDM-UHFFFAOYSA-N | |
| 80008 | |
| CCOC(=O)C1=CN=C(N=C1Cl)SC |
| C8H9ClN2O2S | |
| MFCD00006085 | |
| ethyl 4-chloro-2-methylthio pyrimidine-5-carboxylate, ethyl 4-chloro-2-methylthio-5-pyrimidinecarboxylate, ethyl 4-chloro-2-methylthiopyrimidine-5-carboxylate, 4-chloro-2-methylsulfanyl-pyrimidine-5-carboxylic acid ethyl ester, ethyl 4-chloro-2-methylsulfanyl pyrimidine-5-carboxylate, 2-methylthio-4-chloro-5-ethoxycarbonylpyrimidine, 4-chloro-5-carbethoxy-2-methylthiopyrimidine, 5-pyrimidinecarboxylic acid, 4-chloro-2-methylthio-, ethyl ester, 4-chloro-2-methylthio pyrimidine-5-carboxylic acid ethyl ester, 4-chloro-2-methylsulfanylpyrimidine-5-carboxylic acid ethyl ester | |
| ethyl 4-chloro-2-methylsulfanylpyrimidine-5-carboxylate |
Safety and Handling
TSCA : No