accessibility menu, dialog, popup

UserName

CAS RN 5909-24-0

IUPAC Name: ethyl 4-chloro-2-(methylsulfanyl)pyrimidine-5-carboxylate
Synonyms: ethyl 4-chloro-2-(methylsulfanyl)pyrimidine-5-carboxylate
Molecular Weight (g/mol): 232.68
Molecular Formula: C8H9ClN2O2S
InChi Key: SNNHLSHDDGJVDM-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CN=C(SC)N=C1Cl
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (2)
  • (3)
  • (2)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (4)
  • (2)