missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-Chloro-2-(methylthio)pyrimidine-5-carboxylate 97.0+%, TCI America™

Catalog No. E08415G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E08415G 5 g
E084125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E08415G Supplier TCI America Supplier No. E08415G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5909-24-0
Molecular Formula C8H9ClN2O2S
Molecular Weight (g/mol) 232.682
MDL Number MFCD00006085
InChI Key SNNHLSHDDGJVDM-UHFFFAOYSA-N
Synonym ethyl 4-chloro-2-methylthio pyrimidine-5-carboxylate, ethyl 4-chloro-2-methylthio-5-pyrimidinecarboxylate, ethyl 4-chloro-2-methylthiopyrimidine-5-carboxylate, 4-chloro-2-methylsulfanyl-pyrimidine-5-carboxylic acid ethyl ester, ethyl 4-chloro-2-methylsulfanyl pyrimidine-5-carboxylate, 2-methylthio-4-chloro-5-ethoxycarbonylpyrimidine, 4-chloro-5-carbethoxy-2-methylthiopyrimidine, 5-pyrimidinecarboxylic acid, 4-chloro-2-methylthio-, ethyl ester, 4-chloro-2-methylthio pyrimidine-5-carboxylic acid ethyl ester, 4-chloro-2-methylsulfanylpyrimidine-5-carboxylic acid ethyl ester
PubChem CID 80008
IUPAC Name ethyl 4-chloro-2-methylsulfanylpyrimidine-5-carboxylate
SMILES CCOC(=O)C1=CN=C(N=C1Cl)SC

Specifications

Melting Point 62°C
Color White-Yellow
Boiling Point 132°C
Quantity 5 g
Formula Weight 232.68
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 4-Chloro-2-(methylthio)pyrimidine-5-carboxylate