missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-chloro-2-methylthio-5-pyrimidinecarboxylate, 98%

Catalog No. AC163750250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC163750250 25 g Glass bottle
AC163751000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC163750250 Supplier Thermo Scientific Chemicals Supplier No. 163750250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5909-24-0 | C8H9ClN2O2S | 232.68 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5909-24-0
Molecular Formula C8H9ClN2O2S
Molecular Weight (g/mol) 232.68
MDL Number MFCD00006085
InChI Key SNNHLSHDDGJVDM-UHFFFAOYSA-N
Synonym ethyl 4-chloro-2-methylthio pyrimidine-5-carboxylate, ethyl 4-chloro-2-methylthio-5-pyrimidinecarboxylate, ethyl 4-chloro-2-methylthiopyrimidine-5-carboxylate, 4-chloro-2-methylsulfanyl-pyrimidine-5-carboxylic acid ethyl ester, ethyl 4-chloro-2-methylsulfanyl pyrimidine-5-carboxylate, 2-methylthio-4-chloro-5-ethoxycarbonylpyrimidine, 4-chloro-5-carbethoxy-2-methylthiopyrimidine, 5-pyrimidinecarboxylic acid, 4-chloro-2-methylthio-, ethyl ester, 4-chloro-2-methylthio pyrimidine-5-carboxylic acid ethyl ester, 4-chloro-2-methylsulfanylpyrimidine-5-carboxylic acid ethyl ester
PubChem CID 80008
IUPAC Name ethyl 4-chloro-2-methylsulfanylpyrimidine-5-carboxylate
SMILES CCOC(=O)C1=CN=C(N=C1Cl)SC

Specifications

Melting Point 60.0°C to 63.0°C
Color Beige to White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 25 g
Formula Weight 232.68
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 4-chloro-2-methylthio-5-pyrimidinecarboxylate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 227-619-

RUO – Research Use Only