missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-bromophenylacetate, 98%

Catalog No. AAB2216506
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2216506 5 g
AAB2216514 25 g
AAB2216522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2216506 Supplier Thermo Scientific Chemicals Supplier No. B2216506

CAS: 14062-25-0 | C10H11BrO2 | 243.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14062-25-0
Molecular Formula C10H11BrO2
Molecular Weight (g/mol) 243.1
MDL Number MFCD00016333
InChI Key ZFDCWHPNBWPPHG-UHFFFAOYSA-N
Synonym ethyl 4-bromophenylacetate, ethyl 2-4-bromophenyl acetate, 4-bromophenylacetic acid ethyl ester, benzeneacetic acid, 4-bromo-, ethyl ester, 4-bromo-phenyl-acetic acid ethyl ester, ethyl 4-bromophenyl acetate, 4-bromo-benzeneacetic acid ethyl ester, ethyl-4-bromophenylacetate, pubchem20039, acmc-209ckw
PubChem CID 7020609
IUPAC Name ethyl 2-(4-bromophenyl)acetate
SMILES CCOC(=O)CC1=CC=C(C=C1)Br

Specifications

Melting Point 29°C to 31°C
Boiling Point 88°C to 90°C (0.35 mmHg)
Quantity 5 g
Formula Weight 243.1
Percent Purity 98%
Chemical Name or Material Ethyl 4-bromophenylacetate
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only