missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 4-bromocrotonate, tech. 80%
Description
Ethyl 4-bromocrotonate is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethyl 4-bromocrotonate is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.
Solubility
Insoluble in water.
Notes
Store at 4°C. Incompatible with acids, bases, oxidizing agents, reducing agents. Protect from heat. Store away from strong bases.
Ethyl 4-bromocrotonate is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.
Solubility
Insoluble in water.
Notes
Store at 4°C. Incompatible with acids, bases, oxidizing agents, reducing agents. Protect from heat. Store away from strong bases.
Specifications
Specifications
| Density | 1.402 |
| Boiling Point | 94°C to 95°C (12 mmHg) |
| Flash Point | 97°C (206°F) |
| Refractive Index | 1.493 |
| Quantity | 10 g |
| UN Number | UN1760 |
| Beilstein | 1721388 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 193.05 |
| Percent Purity | 80% |
| Show More |
Safety and Handling
| Hazard Category | H314-H335 |
| Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S. |
| TSCA | No |
| Recommended Storage | Keep cold |
RUO – Research Use Only