Learn More
Ethyl 4-bromocrotonate, 75%, tech.
CAS: 37746-78-4 | C6H9BrO2 | 193.04 g/mol
$68.45 - $594.68
Chemical Identifiers
| CAS | 37746-78-4 |
|---|---|
| Molecular Formula | C6H9BrO2 |
| Molecular Weight (g/mol) | 193.04 |
| MDL Number | MFCD00000247 |
| InChI Key | FHGRPBSDPBRTLS-ONEGZZNKSA-N |
| Synonym | ethyl 4-bromocrotonate, e-ethyl 4-bromobut-2-enoate, ethyl 4-bromobut-2-enoate, 4-bromocrotonic acid ethyl ester, ethyl 2e-4-bromobut-2-enoate, 2-butenoic acid, 4-bromo-, ethyl ester, e, 2-butenoic acid, 4-bromo-, ethyl ester, ethyl e-4-bromo-2-butenoate, ethyl e-4-bromobut-2-enoate, ethyl trans-4-bromo-2-butenoate |
| PubChem CID | 5373944 |
| IUPAC Name | ethyl (E)-4-bromobut-2-enoate |
| SMILES | CCOC(=O)C=CCBr |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC118120050
|
Thermo Scientific Chemicals
118120050 |
5 g | Glass bottle |
Each for $68.45
|
|
||||
|
AC118120250
|
Thermo Scientific Chemicals
118120250 |
25 g | Glass bottle |
Each for $153.44
|
|
||||
|
AC118121000
|
Thermo Scientific Chemicals
118121000 |
100 g | Glass bottle |
Each for $594.68
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 37746-78-4 | |
| 193.04 | |
| FHGRPBSDPBRTLS-ONEGZZNKSA-N | |
| 5373944 | |
| CCOC(=O)C=CCBr |
| C6H9BrO2 | |
| MFCD00000247 | |
| ethyl 4-bromocrotonate, e-ethyl 4-bromobut-2-enoate, ethyl 4-bromobut-2-enoate, 4-bromocrotonic acid ethyl ester, ethyl 2e-4-bromobut-2-enoate, 2-butenoic acid, 4-bromo-, ethyl ester, e, 2-butenoic acid, 4-bromo-, ethyl ester, ethyl e-4-bromo-2-butenoate, ethyl e-4-bromobut-2-enoate, ethyl trans-4-bromo-2-butenoate | |
| ethyl (E)-4-bromobut-2-enoate |
Specifications
| 37746-78-4 | |
| 1.4000g/mL | |
| 97°C | |
| 70% min. (GC) | |
| C6H9BrO2 | |
| BrCH2CH=CHCO2C2H5 | |
| 5 g | |
| ethyl 4-bromocrotonate, e-ethyl 4-bromobut-2-enoate, ethyl 4-bromobut-2-enoate, 4-bromocrotonic acid ethyl ester, ethyl 2e-4-bromobut-2-enoate, 2-butenoic acid, 4-bromo-, ethyl ester, e, 2-butenoic acid, 4-bromo-, ethyl ester, ethyl e-4-bromo-2-butenoate, ethyl e-4-bromobut-2-enoate, ethyl trans-4-bromo-2-butenoate | |
| CCOC(=O)C=CCBr | |
| 193.04 | |
| 193.04 | |
| Technical | |
| Ethyl 4-bromocrotonate, tech., 75% |
| Yellow to Brown | |
| 94°C to 95°C (12.0 mmHg) | |
| Authentic | |
| Glass bottle | |
| 1.4900 to 1.494 | |
| MFCD00000247 | |
| 1.4 | |
| FHGRPBSDPBRTLS-ONEGZZNKSA-N | |
| ethyl (E)-4-bromobut-2-enoate | |
| 5373944 | |
| 75% | |
| Liquid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with
GHS Signal Word: Danger
RUO – Research Use Only