missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-Bromobutyrate 97.0+%, TCI America™

Catalog No. B099925G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B099925G 25 g
B0999100G 100 g
B0999500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B099925G Supplier TCI America Supplier No. B099925G

Chemical Identifiers

CAS 2969-81-5
Molecular Formula C6H11BrO2
Molecular Weight (g/mol) 195.056
MDL Number MFCD00000259
InChI Key XBPOBCXHALHJFP-UHFFFAOYSA-N
Synonym ethyl 4-bromobutyrate, 4-bromobutyric acid ethyl ester, ethyl-4-bromobutyrate, butanoic acid, 4-bromo-, ethyl ester, ethyl-4-bromotbutyrate, 4-bromo-n-butyric acid ethyl ester, brch2ch2ch2c o oc2h5, 4-bromo-butyric acid ethyl ester, 4-bromo-butanoic acid ethyl ester, gamma-bromobutyric acid ethyl ester
PubChem CID 76300
IUPAC Name ethyl 4-bromobutanoate
SMILES CCOC(=O)CCCBr

Specifications

Color Yellow
Boiling Point 200°C
Quantity 25 g
UN Number 3272
Formula Weight 195.06
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 4-Bromobutyrate