missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-bromobutyrate, 97+%

Catalog No. AC173355000
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC173355000 500 mL Glass bottle
AC173351000 100 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC173355000 Supplier Thermo Scientific Chemicals Supplier No. 173355000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2969-81-5 | C6H11BrO2 | 195.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2969-81-5
Molecular Formula C6H11BrO2
Molecular Weight (g/mol) 195.06
InChI Key XBPOBCXHALHJFP-UHFFFAOYSA-N
Synonym ethyl 4-bromobutyrate, 4-bromobutyric acid ethyl ester, ethyl-4-bromobutyrate, butanoic acid, 4-bromo-, ethyl ester, ethyl-4-bromotbutyrate, 4-bromo-n-butyric acid ethyl ester, brch2ch2ch2c o oc2h5, 4-bromo-butyric acid ethyl ester, 4-bromo-butanoic acid ethyl ester, gamma-bromobutyric acid ethyl ester
PubChem CID 76300
IUPAC Name ethyl 4-bromobutanoate
SMILES CCOC(=O)CCCBr

Specifications

Color Colorless to Yellow
Density 1.3600g/mL
Boiling Point 80.0°C to 82.0°C (10.0 mmHg)
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Refractive Index 1.4549 to 1.4569
Linear Formula Br(CH2)3CO2C2H5
Quantity 500 mL
Beilstein 02,283
Specific Gravity 1.36
Solubility Information Solubility in water: insoluble.
Formula Weight 195.06
Percent Purity 97+%
Physical Form Liquid
Chemical Name or Material Ethyl 4-bromobutyrate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN: Wash with plenty of soap a
EINECSNumber 221-005-6

RUO – Research Use Only