missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-Bromo-3-methylbenzoate 98.0+%, TCI America™

Catalog No. p-7129296
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E10365G 5 g
E103625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E10365G Supplier TCI America Supplier No. E10365G

Chemical Identifiers

CAS 160313-69-9
Molecular Formula C10H11BrO2
Molecular Weight (g/mol) 243.10
MDL Number MFCD00673007
InChI Key GIGDAWLJINYIFV-UHFFFAOYSA-N
Synonym 4-Bromo-3-methylbenzoic Acid Ethyl Ester, Ethyl 4-Bromo-m-toluate, 4-Bromo-m-toluic Acid Ethyl Ester
PubChem CID 21729626
IUPAC Name ethyl 4-bromo-3-methylbenzoate
SMILES CCOC(=O)C1=CC(C)=C(Br)C=C1

Specifications

Color Yellow
Boiling Point 275°C
Quantity 5 g
Formula Weight 243.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 4-Bromo-3-methylbenzoate