missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-Aminophenylacetate 98.0+%, TCI America™

Catalog No. A12895G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A12895G 5 g
A128925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A12895G Supplier TCI America Supplier No. A12895G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5438-70-0
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.219
MDL Number MFCD00017569
InChI Key CFNDVXUTYPXOPG-UHFFFAOYSA-N
Synonym ethyl 4-aminophenylacetate, ethyl 2-4-aminophenyl acetate, p-aminophenylacetic acid ethyl ester, 4-aminophenylacetic acid ethyl ester, ethyl 4-aminophenyl acetate, ethyl p-aminophenylacetate, unii-5h8cfu7gnp, benzeneacetic acid, 4-amino-, ethyl ester, ethyl-4-aminophenylacetate
PubChem CID 225219
IUPAC Name ethyl 2-(4-aminophenyl)acetate
SMILES CCOC(=O)CC1=CC=C(C=C1)N

Specifications

Melting Point 50°C
Color White-Yellow
Quantity 5 g
Formula Weight 179.22
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 4-Aminophenylacetate