accessibility menu, dialog, popup

UserName

CAS RN 5438-70-0

IUPAC Name: ethyl 2-(4-aminophenyl)acetate
Synonyms: ethyl 2-(4-aminophenyl)acetate
Molecular Weight (g/mol): 179.22
Molecular Formula: C10H13NO2
InChi Key: CFNDVXUTYPXOPG-UHFFFAOYSA-N
SMILES: CCOC(=O)CC1=CC=C(N)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results