missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-aminophenylacetate, 98%

Catalog No. p-7081520
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1152906 5 g
AAA1152914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1152906 Supplier Thermo Scientific Chemicals Supplier No. A1152906
Only null left

CAS: 5438-70-0 | C10H13NO2 | 179.219 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5438-70-0
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.219
MDL Number MFCD00017569
InChI Key CFNDVXUTYPXOPG-UHFFFAOYSA-N
Synonym ethyl 4-aminophenylacetate, ethyl 2-4-aminophenyl acetate, p-aminophenylacetic acid ethyl ester, 4-aminophenylacetic acid ethyl ester, ethyl 4-aminophenyl acetate, ethyl p-aminophenylacetate, unii-5h8cfu7gnp, benzeneacetic acid, 4-amino-, ethyl ester, ethyl-4-aminophenylacetate
PubChem CID 225219
IUPAC Name ethyl 2-(4-aminophenyl)acetate
SMILES CCOC(=O)CC1=CC=C(C=C1)N

Specifications

Melting Point 46°C to 48°C
Quantity 5 g
Formula Weight 179.22
Percent Purity 98%
Chemical Name or Material Ethyl 4-aminophenylacetate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only