missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-Aminobutyrate Hydrochloride 98.0+%, TCI America™

Catalog No. p-7129224
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E09245G 5 g
E092425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E09245G Supplier TCI America Supplier No. E09245G

Chemical Identifiers

CAS 6937-16-2
Molecular Formula C6H14ClNO2
Molecular Weight (g/mol) 167.633
MDL Number MFCD00012915
InChI Key CXVQSUBJMYZELD-UHFFFAOYSA-N
Synonym ethyl 4-aminobutyrate hydrochloride, ethyl 4-aminobutanoate hydrochloride, 4-aminobutyric acid ethyl ester hydrochloride, ethyl 4-aminobutyrate hcl, butanoic acid, 4-amino-, ethyl ester, hydrochloride, 4-aminobutanoic acid ethyl ester hydrochloride, ethyl 4-aminobutanoate, chloride, 4-ethoxy-4-oxobutyl azanium chloride, h-gaba-oet.hcl, wln: z3vo2 &gh
PubChem CID 12649661
IUPAC Name ethyl 4-aminobutanoate;hydrochloride
SMILES CCOC(=O)CCCN.Cl

Specifications

Melting Point 88°C
Color White
Quantity 5 g
Formula Weight 167.63
Percent Purity ≥98.0% (N,T)
Grade Laboratory
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 4-Aminobutyrate Hydrochloride
RTECSNumber ES6650000
TSCA No