accessibility menu, dialog, popup

UserName

CAS RN 6937-16-2

IUPAC Name: 4-ethoxy-4-oxobutan-1-aminium chloride
Synonyms: 4-ethoxy-4-oxobutan-1-aminium chloride
Molecular Weight (g/mol): 167.63
Molecular Formula: C6H14ClNO2
InChi Key: CXVQSUBJMYZELD-UHFFFAOYSA-N
SMILES: [Cl-].CCOC(=O)CCC[NH3+]
Grade 
  • (2)
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)