missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-Aminobenzoate 99.0+%, TCI America™

Catalog No. p-7116801
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A027125G 25 g
A0271500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A027125G Supplier TCI America Supplier No. A027125G
Only null left

Chemical Identifiers

CAS 94-09-7
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00007892
InChI Key BLFLLBZGZJTVJG-UHFFFAOYSA-N
Synonym benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin
PubChem CID 2337
ChEBI CHEBI:116735
IUPAC Name ethyl 4-aminobenzoate
SMILES CCOC(=O)C1=CC=C(N)C=C1

Specifications

Melting Point 90°C
Color White-Yellow
Boiling Point 310°C
Quantity 25 g
UN Number 3077
Formula Weight 165.19
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 4-Aminobenzoate
EINECSNumber (3)-1443
RTECSNumber DG2450000
TSCA Yes