missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-amino-2-mercaptopyrimidine-5-carboxylate, 97%, Thermo Scientific Chemicals

Catalog No. AAL0712106
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0712106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0712106 Supplier Thermo Scientific Chemicals Supplier No. L0712106

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 774-07-2 | Molecular Formula: C7H9N3O2S | Molecular Weight: 199.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 774-07-2
Molecular Formula C7H9N3O2S
Molecular Weight (g/mol) 199.23
MDL Number MFCD00023227
InChI Key DKTWKRWWQKVQQB-UHFFFAOYSA-N
Synonym ethyl 4-amino-2-mercaptopyrimidine-5-carboxylate, ethyl 6-amino-2-thioxo-1,2-dihydropyrimidine-5-carboxylate, ethyl 4-amino-2-sulfanylpyrimidine-5-carboxylate, ethyl 4-amino-2-mercapto-5-pyrimidinecarboxylate, ethyl 4-amino-2-mercapropyrimidine-5-carboxylate, 5-pyrimidinecarboxylic acid, 4-amino-2-mercapto-, ethyl ester, ethyl 4-amino-2-sulfanyl-5-pyrimidinecarboxylate, ethyl 4-amino-2-sulfanylidene-3h-pyrimidine-5-carboxylate, ethyl 4-amino-2-thioxo-1,2-dihydropyrimidine-5-carboxylate, ethyl 6-amino-2-thioxo-1,2-dihydro-5-pyrimidinecarboxylate
PubChem CID 759149
IUPAC Name ethyl 6-amino-2-sulfanylidene-1H-pyrimidine-5-carboxylate
SMILES CCOC(=O)C1=C(N)NC(=S)N=C1

Specifications

Melting Point ∽260°C (decomposition)
Quantity 5 g
Beilstein 176363
Formula Weight 199.23
Percent Purity 97%
Chemical Name or Material Ethyl 4-amino-2-mercaptopyrimidine-5-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber UV7720000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only