accessibility menu, dialog, popup

UserName

CAS RN 774-07-2

IUPAC Name: ethyl 6-amino-2-sulfanylidene-1,2-dihydropyrimidine-5-carboxylate
Synonyms: ethyl 4-amino-2-sulfanylidene-3H-pyrimidine-5-carboxylate
Molecular Weight (g/mol): 199.23
Molecular Formula: C7H9N3O2S
InChi Key: DKTWKRWWQKVQQB-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(N)NC(=S)N=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results