missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-acetylbutyrate, 97%

CAS: 13984-57-1 | C8H14O3 | 158.197 g/mol

$81.73 - $653.16

Chemical Identifiers

CAS 13984-57-1
Molecular Formula C8H14O3
Molecular Weight (g/mol) 158.197
MDL Number MFCD00009213
InChI Key MGPSIDGTLFKDEY-UHFFFAOYSA-N
Synonym ethyl 4-acetylbutyrate, hexanoic acid, 5-oxo-, ethyl ester, 5-oxohexanoic acid ethyl ester, ethyl 4-acetylbutanoate, ethyl 4-acetyl-butanoate, ethyl 4-acetyl-butyrate, acmc-1c2gr, 5-oxo-hexanoic acid ethyl ester, mgpsidgtlfkdey-uhfffaoysa
PubChem CID 84130
IUPAC Name ethyl 5-oxohexanoate
SMILES CCOC(=O)CCCC(=O)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1784806
View Documents
Thermo Scientific Chemicals
L1784806
5 g
Each for $81.73
Only null left
 
AAL1784814
View Documents
Thermo Scientific Chemicals
L1784814
25 g
Each for $259.49
Only null left
 
AAL1784822
View Documents
Thermo Scientific Chemicals
L1784822
100 g
Each for $653.16
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13984-57-1
158.197
MGPSIDGTLFKDEY-UHFFFAOYSA-N
84130
CCOC(=O)CCCC(=O)C
C8H14O3
MFCD00009213
ethyl 4-acetylbutyrate, hexanoic acid, 5-oxo-, ethyl ester, 5-oxohexanoic acid ethyl ester, ethyl 4-acetylbutanoate, ethyl 4-acetyl-butanoate, ethyl 4-acetyl-butyrate, acmc-1c2gr, 5-oxo-hexanoic acid ethyl ester, mgpsidgtlfkdey-uhfffaoysa
ethyl 5-oxohexanoate

Specifications

13984-57-1
65°C (0.76 mmHg)
C8H14O3
MFCD00009213
1762625
MGPSIDGTLFKDEY-UHFFFAOYSA-N
ethyl 5-oxohexanoate
84130
97%
0.989
69°C (156°F)
1.4275
5 g
ethyl 4-acetylbutyrate, hexanoic acid, 5-oxo-, ethyl ester, 5-oxohexanoic acid ethyl ester, ethyl 4-acetylbutanoate, ethyl 4-acetyl-butanoate, ethyl 4-acetyl-butyrate, acmc-1c2gr, 5-oxo-hexanoic acid ethyl ester, mgpsidgtlfkdey-uhfffaoysa
CCOC(=O)CCCC(=O)C
158.197
158.2
Ethyl 4-acetylbutyrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

P210-P280-P370+P378q-P501c

H227

EINECSNumber : 237-776-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only