missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-acetylbutyrate, 97%

Catalog No. aal1784814
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1784814 25 g
AAL1784806 5 g
AAL1784822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1784814 Supplier Thermo Scientific Chemicals Supplier No. L1784814
Only null left

CAS: 13984-57-1 | C8H14O3 | 158.197 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13984-57-1
Molecular Formula C8H14O3
Molecular Weight (g/mol) 158.197
MDL Number MFCD00009213
InChI Key MGPSIDGTLFKDEY-UHFFFAOYSA-N
Synonym ethyl 4-acetylbutyrate, hexanoic acid, 5-oxo-, ethyl ester, 5-oxohexanoic acid ethyl ester, ethyl 4-acetylbutanoate, ethyl 4-acetyl-butanoate, ethyl 4-acetyl-butyrate, acmc-1c2gr, 5-oxo-hexanoic acid ethyl ester, mgpsidgtlfkdey-uhfffaoysa
PubChem CID 84130
IUPAC Name ethyl 5-oxohexanoate
SMILES CCOC(=O)CCCC(=O)C

Specifications

Density 0.989
Boiling Point 65°C (0.76 mmHg)
Flash Point 69°C (156°F)
Refractive Index 1.4275
Quantity 25 g
Beilstein 1762625
Formula Weight 158.2
Percent Purity 97%
Chemical Name or Material Ethyl 4-acetylbutyrate
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 237-776-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only