missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 4-Acetylbenzoate 97.0+%, TCI America™
$74.56 - $246.05
Chemical Identifiers
| CAS | 38430-55-6 |
|---|---|
| Molecular Formula | C11H12O3 |
| Molecular Weight (g/mol) | 192.21 |
| MDL Number | MFCD00013241 |
| InChI Key | GLOAPLPTWAXAIG-UHFFFAOYSA-N |
| PubChem CID | 600911 |
| IUPAC Name | ethyl 4-acetylbenzoate |
| SMILES | CCOC(=O)C1=CC=C(C=C1)C(C)=O |
Chemical Identifiers
| 38430-55-6 | |
| 192.21 | |
| GLOAPLPTWAXAIG-UHFFFAOYSA-N | |
| ethyl 4-acetylbenzoate |
| C11H12O3 | |
| MFCD00013241 | |
| 600911 | |
| CCOC(=O)C1=CC=C(C=C1)C(C)=O |
Specifications
| 38430-55-6 | |
| White-Yellow | |
| MFCD00013241 | |
| 4-acetylbenzoic acid ethyl ester, benzoic acid, 4-acetyl-, ethyl ester, 4-acetyl-benzoic acid ethyl ester, ethyl4-acetylbenzoate, ethyl 4-acetylbenzate, pubchem18162, ethyl 4-acetyl benzoate, ethyl 4-acetyl-benzoate, acmc-1csm7, ethyl 4-acetylbenzoate | |
| CCOC(=O)C1=CC=C(C=C1)C(C)=O | |
| 192.21 | |
| 192.21 | |
| Crystalline Powder |
| 54°C | |
| C11H12O3 | |
| 1 g | |
| GLOAPLPTWAXAIG-UHFFFAOYSA-N | |
| ethyl 4-acetylbenzoate | |
| 600911 | |
| ≥97.0% (GC) | |
| Ethyl 4-Acetylbenzoate |
Safety and Handling
TSCA : No