accessibility menu, dialog, popup

UserName

H3COOOH3C

CAS RN 38430-55-6

IUPAC Name: ethyl 4-acetylbenzoate
Synonyms: ethyl 4-acetylbenzoate
Molecular Weight (g/mol): 192.21
Molecular Formula: C11H12O3
InChi Key: GLOAPLPTWAXAIG-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=C(C=C1)C(C)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results