missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-acetyl-5-oxohexanoate, 98%

Catalog No. AAL1749614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1749614 25 g
AAL1749606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1749614 Supplier Thermo Scientific Chemicals Supplier No. L1749614

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2832-10-2 | C11H21BrO2 | 265.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2832-10-2
MDL Number MFCD00009211

Specifications

Density 1.067
Boiling Point 153°C to 154°C (19 mmHg)
Flash Point >110°C (230°F)
Molecular Formula C10H16O4
Refractive Index 1.4545
Quantity 25 g
Beilstein 1785189
Synonym ethyl 4,4-diacetylbutyrate, 4-acetyl-5-oxohexanoic acid ethyl ester, 4,4-diacetylbutyric acid ethyl ester, ethyl 4-acetyl-5-oxohexanoate, 4-acetyl-5-oxo-hexanoic acid ethyl ester, hexanoic acid,4-acetyl-5-oxo-, ethyl ester, hexanoic acid, 4-acetyl-5-oxo-, ethyl ester
InChI Key CVPCFJRJUCNUJG-VQHVLOKHSA-N
SMILES CCOC(=O)CCC(C(=O)C)C(=O)C
IUPAC Name ethyl 4-acetyl-5-oxohexanoate
Molecular Weight (g/mol) 200.23
PubChem CID 2733934
Formula Weight 200.24
Percent Purity 98%
Chemical Name or Material Ethyl 4-acetyl-5-oxohexanoate
Show More Show Less
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only