accessibility menu, dialog, popup

UserName

H3COOCH3OHCH3O(E)

CAS RN 2832-10-2

IUPAC Name: ethyl (4E)-4-acetyl-5-hydroxyhex-4-enoate
Synonyms: ethyl (4E)-4-acetyl-5-hydroxyhex-4-enoate
Molecular Weight (g/mol): 200.23
Molecular Formula: C10H16O4
InChi Key: CVPCFJRJUCNUJG-VQHVLOKHSA-N
SMILES: CCOC(=O)CC\C(=C(\C)O)C(C)=O
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)