missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-acetyl-5-oxohexanoate, 98%

Catalog No. AAL1749606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1749606 5 g
AAL1749614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1749606 Supplier Thermo Scientific Chemicals Supplier No. L1749606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2832-10-2 | C11H21BrO2 | 265.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2832-10-2
MDL Number MFCD00009211

Specifications

Density 1.067
Boiling Point 153°C to 154°C (19 mmHg)
Flash Point >110°C (230°F)
Molecular Formula C11H21BrO2
Refractive Index 1.4545
Quantity 5 g
Beilstein 1785189
Synonym undecanoic acid, 11-bromo, 11-bromodecanoic acid, 11-bromo-undecanoic acid, 11-bromoundecanoic acid,tech, 11-bromoundecanoicacid, 11bromoundecanoic acid, 11-bromohendecanoic acid, 11-bromo undecanoic acid, ksc489q1h, 11-bromoundecanoic acid
InChI Key IUDGNRWYNOEIKF-UHFFFAOYSA-N
SMILES C(CCCCCBr)CCCCC(=O)O
IUPAC Name 11-bromoundecanoic acid
Molecular Weight (g/mol) 265.19
PubChem CID 17812
Formula Weight 200.24
Percent Purity 98%
Chemical Name or Material Ethyl 4-acetyl-5-oxohexanoate
Show More Show Less
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only