missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 4-Acetamidophenylacetate 98.0+%, TCI America™
Supplier: TCI America E058525G
Specifications
| Ethyl 4-Acetamidophenylacetate | |
| 78°C | |
| C12H15NO3 | |
| 25 g | |
| OOLUNZSKHFNSCD-UHFFFAOYSA-N | |
| ethyl 2-(4-acetamidophenyl)acetate | |
| 10353455 | |
| ≥98.0% (GC) |
| 13475-17-7 | |
| White | |
| MFCD06797104 | |
| 4-Acetamidophenylacetic Acid Ethyl Ester | |
| CCOC(=O)CC1=CC=C(C=C1)NC(=O)C | |
| 221.256 | |
| 221.26 | |
| Crystalline Powder |
Chemical Identifiers
| 13475-17-7 | |
| 221.256 | |
| OOLUNZSKHFNSCD-UHFFFAOYSA-N | |
| 10353455 | |
| CCOC(=O)CC1=CC=C(C=C1)NC(=O)C |
| C12H15NO3 | |
| MFCD06797104 | |
| 4-Acetamidophenylacetic Acid Ethyl Ester | |
| ethyl 2-(4-acetamidophenyl)acetate |
Safety and Handling
TSCA : No