missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-Acetamidophenylacetate 98.0+%, TCI America™

Catalog No. E05855G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E05855G 5 g
E058525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E05855G Supplier TCI America Supplier No. E05855G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13475-17-7
Molecular Formula C12H15NO3
Molecular Weight (g/mol) 221.256
MDL Number MFCD06797104
InChI Key OOLUNZSKHFNSCD-UHFFFAOYSA-N
Synonym 4-Acetamidophenylacetic Acid Ethyl Ester
PubChem CID 10353455
IUPAC Name ethyl 2-(4-acetamidophenyl)acetate
SMILES CCOC(=O)CC1=CC=C(C=C1)NC(=O)C

Specifications

Melting Point 78°C
Color White
Quantity 5 g
Formula Weight 221.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 4-Acetamidophenylacetate