missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4,6-Dichloronicotinate 98.0+%, TCI America™

Catalog No. E10681G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E10681G 1 g
E10685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E10681G Supplier TCI America Supplier No. E10681G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 40296-46-6 | C8H7Cl2NO2 | 220.049 g/mol

Chemical Identifiers

CAS 40296-46-6
Molecular Formula C8H7Cl2NO2
Molecular Weight (g/mol) 220.049
MDL Number MFCD00173839
InChI Key AAUBVINEXCCXOK-UHFFFAOYSA-N
Synonym ethyl 4,6-dichloronicotinate, 4,6-dichloronicotinic acid ethyl ester, 4,6-dichloropyridine-3-carboxylic acid ethyl ester, 3-pyridinecarboxylic acid, 4,6-dichloro-, ethyl ester, 2,4-dichloro-5-carbethoxypyridine, 4,6-dichloro-3-pyridinecarboxylic acid ethyl ester, 4,6-dichloro-nicotinic acid ethyl ester, 2,4-dichloro-5-ethoxycarbonyl pyridine, ethyl 2,4-dichloropyridine-5-carboxylate, pubchem17888
PubChem CID 600045
IUPAC Name ethyl 4,6-dichloropyridine-3-carboxylate
SMILES CCOC(=O)C1=CN=C(C=C1Cl)Cl

Specifications

Melting Point 33°C
Color White-Yellow
Boiling Point 85°C
Quantity 1 g
Formula Weight 220.05
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Ethyl 4,6-Dichloronicotinate