accessibility menu, dialog, popup

UserName

H3COONClCl

CAS RN 40296-46-6

IUPAC Name: ethyl 4,6-dichloropyridine-3-carboxylate
Synonyms: ethyl 4,6-dichloropyridine-3-carboxylate
Molecular Weight (g/mol): 220.05
Molecular Formula: C8H7Cl2NO2
InChi Key: AAUBVINEXCCXOK-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CN=C(Cl)C=C1Cl
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)