missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4,6-dichloronicotinate, 95%, Thermo Scientific Chemicals

Catalog No. AAH6309203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6309203 1 g
AAH6309206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6309203 Supplier Thermo Scientific Chemicals Supplier No. H6309203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Ethyl 4,6-dichloronicotinate, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as a fine chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 4,6-dichloronicotinate, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as a fine chemical intermediate.

Solubility
Soluble in methanol.

Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 40296-46-6
Molecular Formula C8H7Cl2NO2
Molecular Weight (g/mol) 220.049
MDL Number MFCD00234408
InChI Key AAUBVINEXCCXOK-UHFFFAOYSA-N
Synonym ethyl 4,6-dichloronicotinate, 4,6-dichloronicotinic acid ethyl ester, 4,6-dichloropyridine-3-carboxylic acid ethyl ester, 3-pyridinecarboxylic acid, 4,6-dichloro-, ethyl ester, 2,4-dichloro-5-carbethoxypyridine, 4,6-dichloro-3-pyridinecarboxylic acid ethyl ester, 4,6-dichloro-nicotinic acid ethyl ester, 2,4-dichloro-5-ethoxycarbonyl pyridine, ethyl 2,4-dichloropyridine-5-carboxylate, pubchem17888
PubChem CID 600045
IUPAC Name ethyl 4,6-dichloropyridine-3-carboxylate
SMILES CCOC(=O)C1=CN=C(C=C1Cl)Cl

Specifications

Color Yellow
Quantity 1 g
Solubility Information Soluble in methanol.
Formula Weight 220.05
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Ethyl 4,6-dichloronicotinate
Hazard Category H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only