missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4,4,4-trifluorocrotonate, 98%

Catalog No. AAL1028503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1028503 1 g
AAL1028506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1028503 Supplier Thermo Scientific Chemicals Supplier No. L1028503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 25597-16-4 | C6H7F3O2 | 168.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25597-16-4
Molecular Formula C6H7F3O2
Molecular Weight (g/mol) 168.12
MDL Number MFCD00009903
InChI Key ZKRJCMKLCDWROR-ONEGZZNKSA-N
Synonym ethyl 4,4,4-trifluorocrotonate, ethyl 2e-4,4,4-trifluorobut-2-enoate, ethyl4,4,4-trifluorocrotonate, e-ethyl 4,4,4-trifluorobut-2-enoate, 4,4,4-trifluorocrotonic acid ethyl ester, ethyl e-4,4,4-trifluorobut-2-enoate, ethyl-4,4,4-trifluorocrotonate, ethyl 4,4,4-trifluorobut-2-enoate, 4,4,4-trifluoro-but-2-enoic acid ethyl ester, 2-butenoic acid, 4,4,4-trifluoro-, ethyl ester, 2e
PubChem CID 5371261
IUPAC Name ethyl (E)-4,4,4-trifluorobut-2-enoate
SMILES CCOC(=O)\C=C\C(F)(F)F

Specifications

Density 1.125
Boiling Point 113°C to 115°C
Flash Point 25°C (77°F)
Refractive Index 1.3601
Quantity 1 g
UN Number UN3272
Beilstein 1707262
Formula Weight 168.12
Percent Purity 98%
Chemical Name or Material Ethyl 4,4,4-trifluorocrotonate
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only