missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4,4,4-Trifluorobutyrate 98.0+%, TCI America™

Catalog No. p-7129173
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E08301G 1 g
E08305G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E08301G Supplier TCI America Supplier No. E08301G
Only null left

Chemical Identifiers

CAS 371-26-6
Molecular Formula C6H9F3O2
Molecular Weight (g/mol) 170.131
MDL Number MFCD00041398
InChI Key PSRZMXNNQTWAGB-UHFFFAOYSA-N
PubChem CID 2733273
IUPAC Name ethyl 4,4,4-trifluorobutanoate
SMILES CCOC(=O)CCC(F)(F)F

Specifications

Color Yellow
Boiling Point 127°C
Quantity 1 g
UN Number 3272
Synonym ethyl 4,4,4-trifluorobutyrate, ethyl4,4,4-trifluorobutyrate, 4,4,4-trifluorobutyric acid ethyl ester, 4,4,4-trifluoro-butyric acid ethyl ester, butanoic acid, 4,4,4-trifluoro-, ethyl ester, acmc-1cncd, ethyl-4,4,4-trifluorobutyrate, ethyl 3-trifluoromethylpropionate, butanoic acid,4,4,4-trifluoro-, ethyl ester
Formula Weight 170.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 4,4,4-Trifluorobutyrate