accessibility menu, dialog, popup

UserName

CAS RN 371-26-6

IUPAC Name: ethyl 4,4,4-trifluorobutanoate
Synonyms: ethyl 4,4,4-trifluorobutanoate
Molecular Weight (g/mol): 170.13
Molecular Formula: C6H9F3O2
InChi Key: PSRZMXNNQTWAGB-UHFFFAOYSA-N
SMILES: CCOC(=O)CCC(F)(F)F
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)