missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-(1-Piperazinyl)benzoate 98.0+%, TCI America™

Catalog No. p-7129416
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E12605G 5 g
E126025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E12605G Supplier TCI America Supplier No. E12605G
Only null left

Chemical Identifiers

CAS 80518-57-6
Molecular Formula C13H18N2O2
Molecular Weight (g/mol) 234.30
MDL Number MFCD04973340
InChI Key OQEHTFFLOHTFSB-UHFFFAOYSA-N
Synonym ethyl 4-piperazin-1-yl benzoate, ethyl 4-1-piperazinyl benzoate, 4-piperazin-1-yl-benzoic acid ethyl ester, 1-4-ethoxycarbonylphenyl piperazine, ethyl 4-1-piperazino benzoate, ethyl 4-piperazin-1yl benzoate, benzoic acid, 4-1-piperazinyl-, ethyl ester, 4-piperazin-1-yl benzoic acidethylester, 4-1-piperazinyl benzoic acid ethyl ester
PubChem CID 2761178
IUPAC Name ethyl 4-(piperazin-1-yl)benzoate
SMILES CCOC(=O)C1=CC=C(C=C1)N1CCNCC1

Specifications

Melting Point 102°C
Color Yellow
Quantity 5 g
Formula Weight 234.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 4-(1-Piperazinyl)benzoate