accessibility menu, dialog, popup

UserName

H3COONNH

CAS RN 80518-57-6

IUPAC Name: ethyl 4-(piperazin-1-yl)benzoate
Synonyms: ethyl 4-(piperazin-1-yl)benzoate
Molecular Weight (g/mol): 234.3
Molecular Formula: C13H18N2O2
InChi Key: OQEHTFFLOHTFSB-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=C(C=C1)N1CCNCC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results