missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-tert-butyl-1H-pyrazole-5-carboxylate, 97%

Catalog No. p-7033547
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5067803 1 g
AAH5067806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5067803 Supplier Thermo Scientific Chemicals Supplier No. H5067803

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 916791-97-4 | C10H16N2O2 | 196.25 g/mol

Ethyl 3-tert-butyl-1H-pyrazole-5-carboxylate is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 3-tert-butyl-1H-pyrazole-5-carboxylate is used as pharmaceutical intermediates.

Notes
Store in a cool, dry condition in a well sealed container. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 916791-97-4
Molecular Formula C10H16N2O2
Molecular Weight (g/mol) 196.25
MDL Number MFCD00173782
InChI Key RCXMILPYEKVQLB-UHFFFAOYSA-N
Synonym ethyl 3-tert-butyl-1h-pyrazole-5-carboxylate, 5-tert-butyl-2h-pyrazole-3-carboxylic acid ethyl ester, 5-tert-butyl-2h-pyrazole-3-carboxylic acid ethylester, ethyl 5-tert-butyl-1h-pyrazole-3-carboxylate, ethyl 5-tert-butyl-2h-pyrazole-3-carboxylate, 1h-pyrazole-3-carboxylic acid, 5-1,1-dimethylethyl-, ethyl ester, maybridge3_006722, acmc-1cr14
PubChem CID 2743509
IUPAC Name ethyl 5-tert-butyl-1H-pyrazole-3-carboxylate
SMILES CCOC(=O)C1=NNC(=C1)C(C)(C)C

Specifications

Quantity 1 g
Formula Weight 196.25
Percent Purity 97%
Chemical Name or Material Ethyl 3-tert-butyl-1H-pyrazole-5-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only