missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-Pyridylacetate 98.0+%, TCI America™

Catalog No. E08745G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E08745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E08745G Supplier TCI America Supplier No. E08745G

Chemical Identifiers

CAS 39931-77-6
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
MDL Number MFCD00006411
InChI Key RPWXYCRIAGBAGY-UHFFFAOYSA-N
Synonym ethyl 3-pyridylacetate, ethyl 2-pyridin-3-yl acetate, ethyl 3-pyridineacetate, 3-pyridineacetic acid, ethyl ester, acetic acid, 2-3-pyridyl-, ethyl ester, pyridin-3-yl-acetic acid ethyl ester, 3-pyridylacetic acid ethyl ester, ethyl 2-3-pyridyl acetate, pubchem20506, ethyl-3-pyridyl-acetate
PubChem CID 96490
IUPAC Name ethyl 2-pyridin-3-ylacetate
SMILES CCOC(=O)CC1=CN=CC=C1

Specifications

Boiling Point 120°C
Quantity 5 g
Formula Weight 165.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 3-Pyridylacetate
RTECSNumber UR8950000
TSCA No